2-Chloro-3-amino-4-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-chloro-4-(trifluoromethyl)pyridine

PC520583-10G PC520583-1G PC520583-5G
APOSPC520583-10GEA 1130 EUR
APOSPC520583-10G APOSPC520583-1G APOSPC520583-5G
2-Chloro-3-amino-4-(trifluoromethyl)pyridine 97%
2-Chloro-3-amino-4-(trifluoromethyl)pyridine
Formel: C₆H₄ClF₃N₂
Molecular Weight: 196.557 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09863900
CAS-Nummer: 166770-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR