4-Fluoro-3-(trifluoromethyl)benzenesulphonamide 97%

Lieferant: Apollo Scientific

PC520589-1G PC520589-25G PC520589-5G
APOSPC520589-1GEA 59.6 EUR
APOSPC520589-1G APOSPC520589-25G APOSPC520589-5G
4-Fluoro-3-(trifluoromethyl)benzenesulphonamide 97%
4-Fluoro-3-(trifluoromethyl)benzenesulphonamide
Formel: C₇H₅F₄NO₂S
Molecular Weight: 243,18 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10687234
CAS-Nummer: 1008304-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR