1-Bromo-2-(2,2,2-trifluoroethyl)benzene 97%

Lieferant: Apollo Scientific

PC520594-10G PC520594-1G PC520594-5G
APOSPC520594-10GEA 745 EUR
APOSPC520594-10G APOSPC520594-1G APOSPC520594-5G
1-Bromo-2-(2,2,2-trifluoroethyl)benzene 97%
1-Bromo-2-(2,2,2-trifluoroethyl)benzene
Formel: C₈H₆BrF₃
Molecular Weight: 239.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09996898
CAS-Nummer: 1027513-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR