Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate 97%
Lieferant: Apollo Scientific
Synonyme:
1-(Trifluoromethyl)cyclobutanecarboxylic acid ethyl ester, Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate
PC520596-1G
PC520596-5G
APOSPC520596-1GEA
129
EUR
APOSPC520596-1G
APOSPC520596-5G
Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate 97%
Ethyl-1-(trifluoromethyl)cyclobutanecarboxylate
Formel:
C₈H₁₁F₃O₂ Molecular Weight: 196,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699116 CAS-Nummer: 1040683-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...