2-Amino-5-(trifluoromethyl)nicotinic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-(trifluoromethyl)pyridine-3-carboxylic acid

PC520600-250MG PC520600-500MG
APOSPC520600-250MGEA 280 EUR
APOSPC520600-250MG APOSPC520600-500MG
2-Amino-5-(trifluoromethyl)nicotinic acid 95%
2-Amino-5-(trifluoromethyl)nicotinic acid
Formel: C₇H₅F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10696245
CAS-Nummer: 944900-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR