3-(Difluormethyl)pyridin 97%

Lieferant: Apollo Scientific

PC520604-1G PC520604-5G
APOSPC520604-1GEA 210 EUR
APOSPC520604-1G APOSPC520604-5G
3-(Difluormethyl)pyridin 97%
3-(Difluoromethyl)pyridine
Formel: C₆H₅F₂N
Molecular Weight: 129,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226584
CAS-Nummer: 76541-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR