2-Bromo-3-methylbenzotrifluoride 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-1-methyl-3-(trifluoromethyl)benzene

PC520608-10G PC520608-5G
APOSPC520608-10GEA 616 EUR
APOSPC520608-10G APOSPC520608-5G
2-Bromo-3-methylbenzotrifluoride 95%
2-Bromo-3-methylbenzotrifluoride
Formel: C₈H₆BrF₃
Molecular Weight: 239,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100539
CAS-Nummer: 1099597-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR