3,5-Difluoro-2-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC520612-10G PC520612-1G PC520612-5G
APOSPC520612-10GEA 1060 EUR
APOSPC520612-10G APOSPC520612-1G APOSPC520612-5G
3,5-Difluoro-2-(trifluoromethyl)pyridine 97%
3,5-Difluoro-2-(trifluoromethyl)pyridine
Formel: C₆H₂F₅N
Siedepunkt: 64 °C/150mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD11226519
CAS-Nummer: 1099598-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR