1,3-Dichloro-2-(2,2,2-trifluoroethyl)benzene 95%
Lieferant: Apollo Scientific
PC520616-10G
PC520616-1G
PC520616-5G
APOSPC520616-10GEA
1160
EUR
APOSPC520616-10G
APOSPC520616-1G
APOSPC520616-5G
1,3-Dichloro-2-(2,2,2-trifluoroethyl)benzene 95%
1,3-Dichloro-2-(2,2,2-trifluoroethyl)benzene
Formel:
C₈H₅Cl₂F₃ Molecular Weight: 229.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226581 CAS-Nummer: 1099598-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...