1-Bromo-4-(3,3,3-trifluoropropyl)benzene 95%
Lieferant: Apollo Scientific
PC520619-1G
PC520619-5G
APOSPC520619-1GEA
159
EUR
APOSPC520619-1G
APOSPC520619-5G
1-Bromo-4-(3,3,3-trifluoropropyl)benzene 95%
1-Bromo-4-(3,3,3-trifluoropropyl)benzene
Formel:
C₉H₈BrF₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226575 CAS-Nummer: 1099597-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...