2,4-Difluor-3-nitrobenzonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC520622-1G
PC520622-5G
APOSPC520622-1GEA
164
EUR
APOSPC520622-1G
APOSPC520622-5G
2,4-Difluor-3-nitrobenzonitril 97%
2,4-Difluor-3-nitrobenzonitril
Formel:
C₇H₂F₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226536 CAS-Nummer: 1186194-75-1 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...