1-(2,2,2-Trifluoroethyl)-4-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520628-10G PC520628-1G PC520628-5G
APOSPC520628-10GEA 1280 EUR
APOSPC520628-10G APOSPC520628-1G APOSPC520628-5G
1-(2,2,2-Trifluoroethyl)-4-(trifluoromethyl)benzene 95%
1-(2,2,2-Trifluoroethyl)-4-(trifluoromethyl)benzene
Formel: C₉H₆F₆
Molecular Weight: 228.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226545
CAS-Nummer: 157928-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR