1-Chloro-2-fluoro-3,4-bis-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520632-1G PC520632-5G
APOSPC520632-1GEA 222 EUR
APOSPC520632-1G APOSPC520632-5G
1-Chloro-2-fluoro-3,4-bis-(trifluoromethyl)benzene 95%
1-Chloro-2-fluoro-3,4-bis-(trifluoromethyl)benzene
Formel: C₈H₂ClF₇
Lagertemperatur: Raumtemperatur
MDL: MFCD11226663
CAS-Nummer: 1099597-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR