4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene 95%
Lieferant: Apollo Scientific
PC520633-10G
PC520633-1G
PC520633-5G
APOSPC520633-10GEA
1630
EUR
APOSPC520633-10G
APOSPC520633-1G
APOSPC520633-5G
4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene 95%
4-Chloro-2-(2,2,2-trifluoroethyl)-1-(trifluoromethyl)benzene
Formel:
C₉H₅ClF₆ Molecular Weight: 262.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226668 CAS-Nummer: 1099597-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...