2-Phenyl-4-(trifluoromethyl)quinoline 97%

Lieferant: Apollo Scientific

PC520636-1G PC520636-5G
APOSPC520636-1GEA 176 EUR
APOSPC520636-1G APOSPC520636-5G
2-Phenyl-4-(trifluoromethyl)quinoline 97%
2-Phenyl-4-(trifluoromethyl)quinoline
Formel: C₁₆H₁₀F₃N
Molecular Weight: 273.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226599
CAS-Nummer: 64196-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR