2-Methyl-5-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific

PC520638-10G PC520638-5G
APOSPC520638-10GEA 580 EUR
APOSPC520638-10G APOSPC520638-5G
2-Methyl-5-(trifluoromethyl)pyridine 97%
2-Methyl-5-(trifluoromethyl)pyridine
Formel: C₇H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD11100538
CAS-Nummer: 31181-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR