(2,2,2-Trifluoro-1-methylethyl)benzene 97%

Lieferant: Apollo Scientific

PC520642-1G PC520642-500MG PC520642-5G
APOSPC520642-1GEA 315 EUR
APOSPC520642-1G APOSPC520642-500MG APOSPC520642-5G
(2,2,2-Trifluoro-1-methylethyl)benzene 97%
(2,2,2-Trifluoro-1-methylethyl)benzene
Formel: C₉H₉F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11226603
CAS-Nummer: 161361-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR