3-Chloro-2-methylbenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-2-methyl-3-(trifluoromethyl)benzene

PC520643-1G PC520643-25G PC520643-5G
APOSPC520643-1GEA 72.5 EUR
APOSPC520643-1G APOSPC520643-25G APOSPC520643-5G
3-Chloro-2-methylbenzotrifluoride 97%
3-Chloro-2-methylbenzotrifluoride
Formel: C₈H₆ClF₃
Molecular Weight: 194,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226662
CAS-Nummer: 93242-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR