2-Bromo-1-fluoro-4-(2,2,2-trifluoroethyl)benzene 97%

Lieferant: Apollo Scientific

PC520645-10G PC520645-1G PC520645-5G
APOSPC520645-10GEA 1160 EUR
APOSPC520645-10G APOSPC520645-1G APOSPC520645-5G
2-Bromo-1-fluoro-4-(2,2,2-trifluoroethyl)benzene 97%
2-Bromo-1-fluoro-4-(2,2,2-trifluoroethyl)benzene
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226531
CAS-Nummer: 1099598-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR