2-Chloro-5-(trifluoromethyl)pyridine-3-carbonyl chloride 97%
Lieferant: Apollo Scientific
PC520653-1G
PC520653-5G
APOSPC520653-1GEA
228
EUR
APOSPC520653-1G
APOSPC520653-5G
2-Chloro-5-(trifluoromethyl)pyridine-3-carbonyl chloride 97%
2-Chloro-5-(trifluoromethyl)pyridine-3-carbonyl chloride
Formel:
C₇H₂Cl₂F₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226642 CAS-Nummer: 1099597-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...