1,2,3,4,5-Pentafluoro-6-(2,2,2-trifluoroethyl)benzene 95%

Lieferant: Apollo Scientific

PC520659-1G PC520659-5G
APOSPC520659-1GEA 222 EUR
APOSPC520659-1G APOSPC520659-5G
1,2,3,4,5-Pentafluoro-6-(2,2,2-trifluoroethyl)benzene 95%
1,2,3,4,5-Pentafluoro-6-(2,2,2-trifluoroethyl)benzene
Formel: C₈H₂F₈
Molecular Weight: 250.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226681
CAS-Nummer: 27059-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR