(2,6-Dichloropyrimidin-4-yl)(4-fluorophenyl)methanone 95%

Lieferant: Apollo Scientific

PC520661-1G PC520661-500MG PC520661-5G
APOSPC520661-1GEA 176 EUR
APOSPC520661-1G APOSPC520661-500MG APOSPC520661-5G
(2,6-Dichloropyrimidin-4-yl)(4-fluorophenyl)methanone 95%
(2,6-Dichloropyrimidin-4-yl)(4-fluorophenyl)methanone
Formel: C₁₁H₅Cl₂FN₂O
Molecular Weight: 271.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100517
CAS-Nummer: 1099597-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR