1-Fluoro-2-(2,2,2-trifluoroethyl)benzene 97%
Lieferant: Apollo Scientific
PC520670-10G
PC520670-1G
PC520670-500MG
APOSPC520670-10GEA
1120
EUR
APOSPC520670-10G
APOSPC520670-1G
APOSPC520670-500MG
1-Fluoro-2-(2,2,2-trifluoroethyl)benzene 97%
1-Fluoro-2-(2,2,2-trifluoroethyl)benzene
Formel:
C₈H₆F₄ Molecular Weight: 178.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100172 CAS-Nummer: 93524-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...