4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 97%

Lieferant: Apollo Scientific

PC520672-1G PC520672-5G PC520672-250MG
APOSPC520672-1GEA 148 EUR
APOSPC520672-1G APOSPC520672-5G APOSPC520672-250MG
4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 97%
4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100557
CAS-Nummer: 306960-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR