2-Bromo-3,5,6-trifluoropyridine 95%
Lieferant: Apollo Scientific
PC520676-1G
PC520676-5G
APOSPC520676-1GEA
78.3
EUR
APOSPC520676-1G
APOSPC520676-5G
2-Bromo-3,5,6-trifluoropyridine 95%
2-Bromo-3,5,6-trifluoropyridine
Formel:
C₅HBrF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031583 CAS-Nummer: 1186194-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...