2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
Lieferant: Apollo Scientific
Synonyme: Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, Ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
PC520698-1G PC520698-5G
APOSPC520698-1GEA 151 EUR
APOSPC520698-1G APOSPC520698-5G
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride
Formel: C₅H₉ClF₃NO₂
Molecular Weight: 207,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14525479
CAS-Nummer: 193140-71-5