1-Methyl-4-(2,2,2-trifluoroethyl)benzene 95%

Lieferant: Apollo Scientific

PC520699-1G PC520699-5G
APOSPC520699-1GEA 257 EUR
APOSPC520699-1G APOSPC520699-5G
1-Methyl-4-(2,2,2-trifluoroethyl)benzene 95%
1-Methyl-4-(2,2,2-trifluoroethyl)benzene
Formel: C₉H₉F₃
Molecular Weight: 174.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14525512
CAS-Nummer: 50562-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR