1-Nitro-4-(2,2,2-trifluoroethyl)benzene 95%
Lieferant: Apollo Scientific
PC520701-1G
PC520701-25G
PC520701-5G
APOSPC520701-1GEA
118
EUR
APOSPC520701-1G
APOSPC520701-25G
APOSPC520701-5G
1-Nitro-4-(2,2,2-trifluoroethyl)benzene 95%
1-Nitro-4-(2,2,2-trifluoroethyl)benzene
Formel:
C₈H₆F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14525531 CAS-Nummer: 3764-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...