4-(Difluoromethyl)-2-fluoro-1-methylbenzene 97%
Lieferant: Apollo Scientific
PC520705-1G
PC520705-5G
APOSPC520705-1GEA
164
EUR
APOSPC520705-1G
APOSPC520705-5G
4-(Difluoromethyl)-2-fluoro-1-methylbenzene 97%
4-(Difluoromethyl)-2-fluoro-1-methylbenzene
Formel:
C₈H₇F₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698598 CAS-Nummer: 1214334-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...