1-Chloro-4-(difluoromethyl)-2-fluorobenzene 95%

Lieferant: Apollo Scientific

PC520711-10G PC520711-1G PC520711-5G
APOSPC520711-10GEA 651 EUR
APOSPC520711-10G APOSPC520711-1G APOSPC520711-5G
1-Chloro-4-(difluoromethyl)-2-fluorobenzene 95%
1-Chloro-4-(difluoromethyl)-2-fluorobenzene
Formel: C₇H₄ClF₃
Molecular Weight: 180.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698548
CAS-Nummer: 1214333-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR