1-(Difluoromethyl)-3-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520713-10G PC520713-1G PC520713-5G
APOSPC520713-10GEA 1280 EUR
APOSPC520713-10G APOSPC520713-1G APOSPC520713-5G
1-(Difluoromethyl)-3-(trifluoromethyl)benzene 95%
1-(Difluoromethyl)-3-(trifluoromethyl)benzene
Formel: C₈H₅F₅
Molecular Weight: 196.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698632
CAS-Nummer: 1214358-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR