2-Chlor-8-fluorchinolin 95%

Lieferant: Apollo Scientific

PC520725-25G PC520725-5G
APOSPC520725-25GEA 745 EUR
APOSPC520725-25G APOSPC520725-5G
2-Chlor-8-fluorchinolin 95%
2-Chloro-8-fluoroquinoline
Formel: C₉H₅ClFN
Molecular Weight: 181,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140178
CAS-Nummer: 124467-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR