1-(Trifluoromethyl)cyclopropanemethanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-(Trifluoromethyl)cyclopropyl)methanol

PC520735-1G PC520735-5G
APOSPC520735-1GEA 364 EUR
APOSPC520735-1G APOSPC520735-5G
1-(Trifluoromethyl)cyclopropanemethanol 95%
1-(Trifluoromethyl)cyclopropanemethanol
Formel: C₅H₇F₃O
Molecular Weight: 140,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16990760
CAS-Nummer: 371917-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR