3-Bromo-2-(difluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC520736-1G PC520736-5G
APOSPC520736-1GEA 361 EUR
APOSPC520736-1G APOSPC520736-5G
3-Bromo-2-(difluoromethyl)pyridine 95%
3-Bromo-2-(difluoromethyl)pyridine
Formel: C₆H₄BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD16875638
CAS-Nummer: 1211520-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR