1,1-Difluoro-2-methylcyclohexane 95%
Lieferant: Apollo Scientific
PC520748-1G
PC520748-5G
APOSPC520748-1GEA
209
EUR
APOSPC520748-1G
APOSPC520748-5G
1,1-Difluoro-2-methylcyclohexane 95%
1,1-Difluoro-2-methylcyclohexane
Formel:
C₇H₁₂F₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442248 CAS-Nummer: 74185-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...