2-Fluoro-4-(difluoromethyl)pyridine 97%

Lieferant: Apollo Scientific

PC520749-1G PC520749-500MG PC520749-5G
APOSPC520749-1GEA 338 EUR
APOSPC520749-1G APOSPC520749-500MG APOSPC520749-5G
2-Fluoro-4-(difluoromethyl)pyridine 97%
2-Fluoro-4-(difluoromethyl)pyridine
Formel: C₆H₄F₃N
Molecular Weight: 147.1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442245
CAS-Nummer: 84940-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR