3-Bromo-2-chloro-5-(difluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
PC520758-1G
PC520758-500MG
PC520758-5G
APOSPC520758-1GEA
372
EUR
APOSPC520758-1G
APOSPC520758-500MG
APOSPC520758-5G
3-Bromo-2-chloro-5-(difluoromethyl)pyridine 97%
3-Bromo-2-chloro-5-(difluoromethyl)pyridine
Formel:
C₆H₃BrClF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199311 CAS-Nummer: 71702-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...