5-Fluor-2-formylphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Fluoro-2-formylphenyl)boronic acid
PC52076-1G
PC52076-25G
PC52076-5G
APOSPC52076-1GEA
21.9
EUR
APOSPC52076-1G
APOSPC52076-25G
APOSPC52076-5G
5-Fluor-2-formylphenylboronsäure
5-Fluor-2-formylphenylboronsäure
Formel:
C₇H₆BFO₃ Molecular Weight: 167,9300 g/mol Schmelzpunkt: 137…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09953731 CAS-Nummer: 1256355-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...