2,2,2-Trifluoro-1-phenylethylamine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2,2,2-Trifluoro-1-phenylethylamine Hydrochloride

PC520790-10G PC520790-1G PC520790-5G
APOSPC520790-10GEA 989 EUR
APOSPC520790-10G APOSPC520790-1G APOSPC520790-5G
2,2,2-Trifluoro-1-phenylethylamine hydrochloride 95%
2,2,2-Trifluoro-1-phenylethylamine hydrochloride
Formel: C₈H₉ClF₃N
Molecular Weight: 211,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00971700
CAS-Nummer: 13652-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR