2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine 98%

Lieferant: Apollo Scientific

PC520791-10G PC520791-1G PC520791-5G
APOSPC520791-10GEA 815 EUR
APOSPC520791-10G APOSPC520791-1G APOSPC520791-5G
2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine 98%
2,6-Dichloro-5-fluoro-3-(hydroxymethyl)pyridine
Formel: C₆H₄Cl₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04972782
CAS-Nummer: 820224-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR