4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 97%

Lieferant: Apollo Scientific

PC520795-1G PC520795-5G
APOSPC520795-1GEA 292 EUR
APOSPC520795-1G APOSPC520795-5G
4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 97%
4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene
Formel: C₈H₅ClF₄
Molecular Weight: 212.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226523
CAS-Nummer: 1099598-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR