2-Methyl-1-nitro-4-(2,2,2-trifluoroethyl)benzene 95%

Lieferant: Apollo Scientific

PC520802-1G PC520802-5G
APOSPC520802-1GEA 361 EUR
APOSPC520802-1G APOSPC520802-5G
2-Methyl-1-nitro-4-(2,2,2-trifluoroethyl)benzene 95%
2-Methyl-1-nitro-4-(2,2,2-trifluoroethyl)benzene
Formel: C₉H₈F₃NO₂
Molecular Weight: 219.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566378
CAS-Nummer: 1404193-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR