4-Fluoro-1H-imidazole 95%

Lieferant: Apollo Scientific

PC520808-100MG PC520808-250MG PC520808-500MG PC520808-1G
APOSPC520808-100MGEA 0 EUR
APOSPC520808-100MG APOSPC520808-250MG APOSPC520808-500MG APOSPC520808-1G
4-Fluoro-1H-imidazole 95%
4-Fluoro-1H-imidazole
Formel: C₃H₃FN₂
Molecular Weight: 86.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835574
CAS-Nummer: 30086-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR