5-(Trifluoromethyl)isoquinoline 97%

Lieferant: Apollo Scientific

PC520907-1G PC520907-5G
APOSPC520907-1GEA 514 EUR
APOSPC520907-1G APOSPC520907-5G
5-(Trifluoromethyl)isoquinoline 97%
5-(Trifluoromethyl)isoquinoline
Formel: C₁₀H₆F₃N
Molecular Weight: 197.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100536
CAS-Nummer: 120568-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR