Methyl-3-fluorpicolinat 97%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-pyridine-2-carboxylic acid methyl ester, Methyl 3-fluoropyridine-2-carboxylate

PC520911-5G
APOSPC520911-5GEA 97.3 EUR
APOSPC520911-5G
Methyl-3-fluorpicolinat 97%
Methyl-3-fluorpicolinat
Formel: C₇H₆FNO₂
Molecular Weight: 155,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659496
CAS-Nummer: 869108-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR