1,1-Dichlordifluorethylen 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1-Dichlorodifluoroethylene
PC52110-100G
PC52110-25G
PC52110-500G
APOSPC52110-100GEA
249
EUR
APOSPC52110-100G
APOSPC52110-25G
APOSPC52110-500G
1,1-Dichlordifluorethylen 90%
1,1-Dichlordifluorethylen
Formel:
C₂Cl₂F₂ Molecular Weight: 132,92 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039338 CAS-Nummer: 79-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...