3,4,5,6-Tetrafluorsalicylsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC52121-1G
PC52121-5G
APOSPC52121-1GEA
154
EUR
APOSPC52121-1G
APOSPC52121-5G
3,4,5,6-Tetrafluorsalicylsäure
3,4,5,6-Tetrafluorsalicylsäure
Formel:
C₇H₂F₄O₃ Schmelzpunkt: 173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483242 CAS-Nummer: 14742-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...