2-Fluor-α-methyl-4-biphenylessigsäure 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
PC5245-100G
PC5245-25G
PC5245-5G
APOSPC5245-100GEA
509
EUR
APOSPC5245-100G
APOSPC5245-25G
APOSPC5245-5G
2-Fluor-α-methyl-4-biphenylessigsäure 98%
2-Fluor-α-methyl-4-biphenylessigsäure
Formel:
C₁₅H₁₃FO₂ Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079303 CAS-Nummer: 5104-49-4 EINECS: 225-827-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...