3-Methoxy-4-(trifluoromethyl)benzylamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-[3-Methoxy-4-(trifluoromethyl)phenyl]methanamine, Benzenemethanamine, 3-methoxy-4-(trifluoromethyl)-

PC52466-1G PC52466-250MG
APOSPC52466-1GEA 799 EUR
APOSPC52466-1G APOSPC52466-250MG
3-Methoxy-4-(trifluoromethyl)benzylamine 95%
3-Methoxy-4-(trifluoromethyl)benzylamine
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398593
CAS-Nummer: 1261854-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR