2-Brom-3-(trifluormethyl)benzonitril 99%

Lieferant: Apollo Scientific
Achtung

PC5271-1G PC5271-5G
APOSPC5271-1GEA 72.5 EUR
APOSPC5271-1G APOSPC5271-5G
2-Brom-3-(trifluormethyl)benzonitril 99%
2-Brom-3-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Molecular Weight: 250,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08532491
CAS-Nummer: 914637-07-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR